missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ IEF Sample Buffer pH 3-10 (2X)

Product Code. 11539076
Click to view available options
Quantity:
25 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11539076

Brand: Invitrogen™ LC5311

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

An optimized pre-mixed IEF sample buffer

Novex IEF Sample Buffer pH 3-10 (2X) is optimized pre-mixed IEF sample buffer for using with Novex pH 3-10 IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.

Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Buffer Sample Loading Buffers
Concentration 2 X
Gel Type IEF
pH Range 3 to 10
Product Type IEF Sample Buffer
Gel Compatibility Novex™ IEF Gels
Product Line Novex
Quantity 25 mL
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.