Learn More
Invitrogen™ IL-1 beta Human ELISA Kit
Quantifies natural and recombinant human IL-1β in serum, buffered solution, or cell culture medium
£477.00 - £1412.00
Specifications
Accession Number | P01584 |
---|---|
Assay Range | 3.9-250 pg/mL |
Assay Sensitivity | 1 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10130573
|
Invitrogen™
KHC0011 |
96 Tests |
£477.00
96 tests |
In Stock
Log in to see stock. |
|||||
10150573
|
Invitrogen™
KHC0012 |
2 x 96 Tests |
£618.00
Each |
In Stock
Log in to see stock. |
|||||
10140573
|
Invitrogen™
KHC0011C |
5 x 96 Tests |
£1412.00
Pack of 5 |
In Stock
Log in to see stock. |
|||||
Description
The Human Interleukin-1beta (Hu IL-1beta) ELISA quantitates Hu IL-1β in human serum, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-1β. Principle of the method The Human IL-1β solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL-1β is important in immune response.
Specifications
P01584 | |
1 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
IL-1,IL1-BETA,IL1F2 | |
4.4% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
3.9-250 pg/mL | |
Biotin | |
Serum, Supernatant | |
Colorimetric | |
3553 | |
6.6% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
1pg/mL | |
Serum, 50 μL; Supernatant, 50 μL | |
Human | |
2 hr. 55 min. |