Learn More
Invitrogen™ IL-6 Mouse ELISA Kit
To detect and quantify the level of mouse IL-6 in serum, plasma, and supernatant using 96-well plates and a microplate reader
£477.00 - £1412.00
Specifications
Accession Number | P08505 |
---|---|
Assay Range | 7.8-500 pg/mL |
Assay Sensitivity | <3 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Includes: Mouse IL-6 Antibody Coated 96-Well Plate, Mouse IL-6 Standard, Standard Diluent Buffer, Mouse IL-6 Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer Concentrate (25X), Stabilized Chromogen, TMB, Stop Solution, Plate Covers, Detailed protocol with validation tests
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10707833
|
Invitrogen™
KMC0061 |
96 Tests |
£477.00
Each |
In Stock
Log in to see stock. |
|||||
10110623
|
Invitrogen™
KMC0062 |
2 x 96 Tests |
£618.00
Each |
In Stock
Log in to see stock. |
|||||
10476283
|
Invitrogen™
KMC0061C |
5 x 96 Tests |
£1412.00
Each |
In Stock
Log in to see stock. |
|||||
Description
The Mouse Interleukin (IL-6)) ELISA quantitates Ms IL-6 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-6. Principle of the method The Mouse IL-6 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
- Easy to run
- Rapid protocol
- Convenient precoated, removable 8-well strips
- Easy-to-Run Sandwich ELISA
- The total assay incubation time is only 3.5 hours.
- A monoclonal capture antibody specific for Mouse IL-6 has been coated onto the wells of the 96-well plate provided
- Samples, including standard of known Mouse IL-6 content, controls, and°Cs, are pipetted into these wells followed by an addition of a biotinylated polyclonal detector antibody
- During the first incubation, Mouse IL-6 antigen binds to the immobilized capture antibody and to solution phase biotinylated detector antibody on second site
- After washing, streptavidin-horseradish peroxidase (HRP) is added
- This binds to the biotinylated detection antibody to complete four-member sandwich
- After second incubation and washing to remove all unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color
- The intensity of this colored product is directly proportional to the concentration of Mouse IL-6 present in the original specimen and the optical density can be read on a standard microplate reader
Convenient Kit Format:The kit comes with a capture antibody precoated to the plate, eliminating an overnight coating process. The plates are machine coated to provide low variability well-to-well. Furthermore, the plate comes as removable 8-well strips allowing you to run as few samples as you want. Each ELISA kit is validated for sensitivity, specificity, precision, and lot-to-lot consistency.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet ice
Specifications
P08505 | |
<3 pg/mL | |
ELISA Kit | |
Mouse | |
Colorimetric Microplate Reader | |
IL-6 | |
5.8% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
7.8-500 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Colorimetric | |
16193 | |
6.7% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
<3pg/mL | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
Mouse | |
3 hr. 30 min. |
For Research Use Only. Not for use in diagnostic procedures.