Learn More
Invitrogen™ IL1RA Human Matched Antibody Pair
Matched Antibody Pair
Brand:  Invitrogen™ CHC1183
Product Code. 10083293
  • £392.00 / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Includes: Antibody Pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Streptavidin-HRP concentrate and technical data sheet.
Requires : Buffer reagents required to complete reaction available separately (Cat. No. CNB0011).
Description
Description
The Human IL-1RA; Antibody Pair Kit comes with pre-matched antibody pairs, standards, and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-1RA. Buffer reagents needed to complete the ELISA reaction are sold separate as a Buffer Kit for Antibody Pairs (Catalog No. CNB0011). The Assay Buffer included in this Buffer Kit can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Interleukin 1 receptor antagonist (ILRA) is a protein related to the IL-1 family based on its similarities in amino acid sequence to both IL-1 beta and IL-1 alpha, similarities in gene structure, and collective gene localization. The human gene contains four exons and maps to chromosome 2q13-14. IL-1 RA binds to the IL-1 receptor and blocks IL-1 action through competitive inhibition. In contrast to IL-1, the binding of IL-1RA does not initiate signaling events. The secretion of IL-1 RA is mediated by a classical signal sequence while neither IL-1alpha nor IL-1beta possess such a sequence. The lack of homology between IL-1 RA and IL-1 alpha or IL-1 beta near the N terminus may provide an explanation for the biosynthetic and functional differences between IL-1 RA and the two IL-1 proteins. The production of IL-1RA is stimulated by many substances including adherent IgG, cytokines, and bacterial or viral components. IL-1RA is an important natural anti-inflammatory protein that when missing or just available in inadequate amounts plays a key role in several pathological conditions such as rheumatoid arthritis, colitis, osteomyelitis, periostitis and pustulosis.
Order Info
Shipping Condition: Wet Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P18510
IL1RN
3557
Human
ELISA Kit
DIRA, ICIL-1RA, IL-1RN, IL-1ra, IL-1ra3, IL1F3, IL1RA, IRAP, MGC10430, MVCD4
10 x 96 Tests
Capture (coating) Antibody, Detection Antibody, Standard, Streptavidin-HRP
2°C to 8°C
Colorimetric Microplate Reader