Learn More
Invitrogen™ ELISA Stop Solution
A ready-to-use solution intended for use as a stop solution in ELISA applications that use horseradish peroxidase and TMB substrate solution.
£74.75 - £202.00
Specifications
Product Type | ELISA Stop Solution |
---|---|
Form | Liquid |
Substrate Type | HRP, TMB |
Content And Storage | Store at 2°C to 8°C |
Shipping Condition | Ambient |
Description
ELISA Stop Solution is a ready-to-use solution intended for use as a stop solution in ELISA applications that use horseradish peroxidase and TMB substrate solution. As TMB substrate solution reacts with horseradish peroxidase, it produces a pronounced blue color. At the end of TMB incubation, a volume of ELISA Stop Solution equal to the volume of TMB substrate solution is added to the wells to terminate the horseradish peroxidase reaction. Addition of Stop Solution results in the rapid formation of a yellow end product. The optical density of this yellow end product is measured at 450 nm. The yellow end product is stable for sixty minutes after addition of Stop Solution and so optical density should be measured within that time.
For a complete kit of ELISA buffers, see ELISA Buffer Kit.
View all available buffers and reagents for ELISA applications.
Order Info
Shipping Condition: Room Temperature
Specifications
ELISA Stop Solution | |
HRP, TMB | |
Ambient |
Liquid | |
Store at 2°C to 8°C |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.