missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Large Cellophane

Product Code. 10789243
Click to view available options
Quantity:
100/ pkg.
Unit Size:
Pack of 100
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10789243

Brand: Invitrogen™ NC2200

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Component of Large Gel Drying Kit for drying PAGE gels

The Large Gel Drying Kit provides a fast and easy way to consistently dry gels without cracking and can accommedate up to 1 to 4 mini-gel(s) or a large format gel.

Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 100/pack; Store at room temperature
Product Type Large Cellophane
Product Line Novex
Quantity 100/ pkg.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.