Learn More
Invitrogen™ Wash Solution Concentrate (20X), 15 ml
Designed to meet additional solution volume for protocols on automated systems
Brand: Invitrogen™ WB04
Code : 50
Additional Details : Weight : 0.02000kg
Includes: Wash Solution Concentrate contains: 15mL of 20X wash solution
Description
For use with Luminex multiplex panels. 20X concentrate contains 0.1% sodium azide.
Please refer to the manualfor details on use of wash solution concentrate
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Specifications
Wash Buffer | |
Liquid | |
MAGPIX™ | |
Wet Ice |
15 mL | |
Luminex Instruments, FlexMAP 3D System, MAGPIX System, Luminex 200 System | |
Wash Solution Concentrate contains:15 mL of 20X wash solution |
For Research Use Only. Not for use in diagnostic procedures.