Learn More
Invitrogen™ Mark12™ Unstained Standard
Description
Includes
1.0 mL (200 applications of 5μL each) provided in a plastic vial Loading Buffer consists of Tris-HCl, Glycerol, SDS, Phenol Red, and Coomassie Blue G-250
Mark12 Unstained Standard is supplied ready-to-use with either NuPAGE™, tris-glycine, or tricine gels. Coomassie™, silver, or other protein staining methods are required to visualize this standard.
- 12 polypeptides resolved into sharp, tight bands in the range of 2.5 to 200kDa
- Ideal molecular weight estimation
- A ready-to-use format no mixing or reducing required
- Gel Compatibility: Bolt Bis-Tris Plus Gels, NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels, NuPAGE Tris-Acetate Gels, SDS-PAGE Gels
This standard is pre-reduced and does not contain reducing reagents in the loading buffer.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Content And Storage | 1.0 mL (200 applications of 5 μL each) provided in a plastic vial. Loading Buffer consists of Tris-HCl, Glycerol, SDS, Phenol Red, and Coomassie™ Blue G-250. Store at 4°C. |
| Number of Markers | 12 |
| Product Type | Protein Ladder |
| Ready to Load | Yes |
| Size Range | 2.5 to 200 kDa |
| Stain Type | Unstained |
| Gel Compatibility | Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, Novex™Tricine, SDS-PAGE Gels |
| Molecular Weight (g/mol) | 200, 116.3, 97.4, 66.3, 55.4, 36.5, 31, 21.5, 14.4, 6, 3.5, 2.5 kDa |
| Product Line | Mark12 |
| Quantity | 1 mL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.