Learn More
Invitrogen™ NativeMark™ Unstained Protein Standard
Ready-to-use protein standard designed for molecular weight estimation of proteins using native gel electrophoresis
Brand: Invitrogen™ LC0725
189.65 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Includes: Supplied in storage buffer (Bis/Tris-HCl, pH 7.0; NaCl; glycerol; Ponceau S)
Description
Electrophoresis under native (nondenaturing) conditions maintains the native protein conformation, cohesion of subunits, and biological activity. During native electrophoresis, proteins are separated based on their charge to mass ratios.
- Versatile: recommended for use with Novex NativePAGE™ Bis-Tris, gradient Tris-Glycine, or NuPAGE Novex Tris-Acetate gels
- Comprehensive: contains markers with a wide range of high molecular weights, providing 8 protein bands in the range of 20–1,200 kDa
- Convenient: no need to add sample buffer, just load and run
- Compatible: can be visualized using Coomassie™, silver, or fluorescent stains after electrophoresis, or with Ponceau S, Coomassie, or other membrane stains after Western transfer
- Gel Compatibility: NativePAGE Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels, NuPAGE Tris-Acetate Gels
1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Dry Ice
Specifications
The NativeMark™ Unstained Protein Standard is supplied in storage buffer (Bis/Tris-HCl, pH 7.0; NaCl; glycerol; Ponceau S). Store at -20°C. |
|
Protein Ladder | |
Native gels | |
Unstained | |
1236, 1048, 720, 480, 242, 146, 66, 20 kDa | |
5 x 50 μL | |
Native-PAGE |
8 | |
Yes | |
20 to 1,200 kDa | |
Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, Novex™Tris-Glycine Gels, NativePAGE™ Gels, NuPAGE™ Tris-Acetate Gels | |
NativeMARK™ | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.