missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NativePAGE™ 5% G-250 Sample Additive
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
Description
This Additive is also included in the NativePAGE Sample Prep Kit. The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE Sample Prep Kit are compatible with NativePAGE Novex Bis-Tris Gels for native electrophoresis showing increased resolution and reduced streaking.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store at room temperature |
| Buffer | Sample Loading Buffers |
| Gel Type | Native-PAGE |
| Product Type | PAGE G-250 Sample Additive |
| Gel Compatibility | NativePAGE™ Gels |
| Product Line | NativePAGE |
| Quantity | 0.5 mL |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction