missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NativePAGE™ Cathode Buffer Additive (20X)

Product Code. 10779663
Click to view available options
Quantity:
250 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10779663

Brand: Invitrogen™ BN2002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

For running NativePAGE Gels

NativePAGE Cathode Buffer Additive (20X) is added to the NativePAGE Running Buffer and the resulting buffer is used in the cathode chamber of an XCell SureLock™ Mini Cell for running NativePAGE Gels.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Buffer Anode & Cathode Buffers
Concentration 20 X
Gel Type IEF
Product Type PAGE Cathode Buffer Additive
Gel Compatibility NativePAGE™ Gels
Product Line NativePAGE
Quantity 250 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.