Learn More
Invitrogen™ NativePAGE™ Running Buffer Kit
Contains the NativePAGE Running Buffer (20X)
Brand: Invitrogen™ BN2007
212.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 1.06000kg
Includes: 1L of NativePAGE Running Buffer (20X), 250mL NativePAGE Cathode Buffer Additive (20X)
Description
NativePAGE Running Buffer (20X), is used to make the NativePAGE Cathode and Anode Running Buffers for use with an XCell SureLock™ Mini Cell when running NativePAGE Gels. The kit also contains the NativePAGECathode Buffer Additive (20X) that is used with the NativePAGE Running Buffer to make the Cathode Running Buffer for the system.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Contains: • 1 L of NativePAGE™ Running Buffer (20X) • 250 ml NativePAGE™ Cathode Buffer Additive (20X) Store at room temperature. |
|
Native-PAGE | |
NativePAGE™ Gels |
Running Buffers | |
PAGE Running Buffer Kit | |
NativePAGE™ |
Safety and Handling
- NativePAGE Running Buffer Kit
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)-1,3-Propanediol
For Research Use Only. Not for use in diagnostic procedures.