missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NativePAGE™ Sample Prep Kit

Includes sample preparation reagents for native gel electrophoresis

Brand:  Invitrogen™ BN2008

Product Code. 10165392

  • £239.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: 1mL of 10% DDM, 1mL of 5% Digitonin, 10mL of NativePAGE, 4X Sample Buffer, 0.5mL NativePAGE 5% G-250 Sample Additive
This item is not returnable. View return policy

Description

Description

The kit includes two ready-to-use detergent solutions, 10% DDM (n-dodecyl-β-D-maltoside) and 5% Digitonin, which improve the solubility of hydrophobic and membrane proteins during sample preparation. The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE Sample Prep Kit are compatible with NativePAGE Novex Bis-Tris Gels for native electrophoresis, showing increased resolution and reduced streaking.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 1 ml of 10% DDM
• 1 ml of 5% Digitonin
• 10 ml of NativePAGE™ 4X Sample Buffer
• 0.5 ml NativePAGE™ 5% G-250 Sample Additive.

Store the kit at 2°C to 8°C.
Native-PAGE
NativePAGE™ Gels
1 kit
Sample Loading Buffers
PAGE Sample Prep Kit
NativePAGE™
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3016198 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberBN2008
Lot Number3050930 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberBN2008
Lot Number2474226 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberBN2008
Lot Number2512482 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberBN2008
Lot Number2474227 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberBN2008
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ NativePAGE™ Sample Prep Kit > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.