Learn More
Invitrogen™ Ni-NTA Purification System
Designed for the purification of recombinant proteins that contain a polyhistidine (6xHis) sequence
Brand: Invitrogen™ K95001
Code : 50
Additional Details : CAS Number : 53123-88-9 Weight : 0.00525kg Transport : UN number : 1170 Chem class : 3 Pack group : III
Includes: 6 x 2mL resin columns and buffers for native and denaturing purification; 10mL Ni-NTA Agarose.
Description
- Utilizes Ni-NTA nickel-chelating resin and is supplied with native and denaturing buffers for efficient purification of recombinant proteins under different conditions
His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Specifications
Suspension | |
Ni-NTA Purification System | |
Six 2-ml resin columns and buffers for native and denaturing purification. Ten milliliters of Ni-NTA Agarose. Store at +4°C. All reagents are guaranteed stable for 6 months when properly stored. | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.