missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NuPAGE™ Antioxidant
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Description
Includes
Contains 15mL NuPAGE Antioxidant
Add to running buffer in the upper (cathode) buffer chamber when performing protein gel electrophoresis under reducing conditions.
- Use with samples reduced using NuPAGE Sample Reducing Agent or other reducing agents
- Optimized for neutral pH of NuPAGE Novex™ gels and is not compatible with other gel chemistries
- May also be added to NuPAGE Transfer Buffer to enhance transfer of proteins to membranes
- Migrates with reduced proteins to maintain reducing conditions and to prevent reoxidation of sensitive amino acids such as methionine and tryptophan
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Content And Storage | Contains 15 ml NuPAGE™ Antioxidant. Store at 4°C. Guaranteed stable for 6 months unless otherwise specified in the product literature. |
| Product Type | Antioxidant |
| Product Line | NuPAGE |
| Quantity | 15 mL |
| Shipping Condition | Wet Ice |
Safety and Handling
Product Identifier
- NuPAGE Antioxidant
- Danger
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H315-Causes skin irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Dimethylformamide
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction