missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ LDS Sample Buffer (4X)

Product Code. p-4918335
Click to view available options
Quantity:
10mL
250mL
Unit Size:
10mL
250mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11549166

Brand: Invitrogen™ NP0007

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

NuPAGE LDS Sample Buffer (4X), 10mL

Used to prepare protein samples for denaturing gel electrophoresis with NuPAGE™ Novex™ gels. Invitrogen™ Novex™ NuPAGE™ LDS Sample Buffer (4X) contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of the reducing agent.

Optimal protein sample preparation for polyacrylamide gel electrophoresis (PAGE) requires denaturing and reducing protein disulfide bonds.

  • Highly viscous and concentrated solution that contains higher a concentration of glycerol
  • Contains twice the amount of LDS compared to the amount of SDS in Novex™ Tris-Glycine SDS Sample Buffer or in Tricine SDS Sample Buffer
  • NuPAGE™ LDS Sample Buffer should be brought to room temperature (25°C) prior to use

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.

Store at 4°C to 25°C.

Buffer Sample Loading Buffers
Concentration 4 X
Gel Type NuPAGE
Product Type LDS Sample Buffer
Gel Compatibility NuPAGE™ Gels
Product Line NuPAGE
Quantity 10mL
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.