missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels)
All buffers are made with high-purity reagents and are strictly quality controlled
Brand: Invitrogen™ NP0050
61.20 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Includes: NuPAGE MOPS SDS Running Buffer (20X, 500mL), NuPAGE Sample Reducing Agent (10X, 250 μL) containing 500mM Dithiothreitol, NuPAGE Antioxidant (15mL), NuPAGE LDS Sample Buffer (4X, 10mL)
Requires : The buffers are provided as a concentrated solution and require simple dilution with deionized water before use.
This item is not returnable. View return policy
Description
The NuPAGE MOPS SDS Buffer Kit, designed for separation of medium- to large-size proteins on NuPAGE Novex Bis-Tris gels. Pre-mixed NuPAGE buffers are a convenient way to ensure high-quality, consistent electrophoresis results.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Running Buffer | |
SDS PAGE Gel | |
NuPAGE Bis-Tris Gel | |
1 kit |
Contains • NuPAGE™ MOPS SDS Running Buffer (20X, 500 ml) • NuPAGE™ Sample Reducing Agent (10X, 250 μl) containing 500 mM Dithiothreitol • NuPAGE™ Antioxidant (15 ml) • NuPAGE™ LDS Sample Buffer (10 ml) Store each at 4°C. Warm NuPAGE™ LDS Sample Buffer to 25°C before use. |
|
Room Temperature | |
NuPAGE™ |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- NuPAGE MOPS SDS Buffer Kit (for Bis-Tris Gels)
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H360-May damage fertility or the unborn child.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Dimethylformamide
For Research Use Only. Not for use in diagnostic procedures.