missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NuPAGE™ MOPS SDS Running Buffer (20X)
Made with high-purity reagents and are strictly quality controlled.
£80.00 - £602.00
Specifications
Chemical Name or Material | Running Buffer |
---|---|
Concentration | 20X |
pH | 7.7 |
Physical Form | Liquid |
Product Line | NuPAGE |
Includes: NuPAGE MOPS SDS Running Buffer (20X)
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11589156
|
Invitrogen™
NP0001 |
500 mL |
£80.00
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11599156
|
Invitrogen™
NP000102 |
5 L |
£602.00
5 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NuPAGE MOPS SDS Running Buffer (20X) is formulated for running proteins on NuPAGE Novex Bis-Tris gels only. NuPAGE™ MOPS SDS Running Buffer is recommended for separating small- to medium-sized proteins.
- Get consistent, convenient, high-quality results
- Pre-mixed NuPAGE buffers are convenient way to ensure high-quality, consistent electrophoresis results
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Informations sur la commande
Shipping Condition: Room Temperature
Spécification
Running Buffer | |
7.7 | |
NuPAGE |
20X | |
Liquid | |
50 mM MOPS, 50 mM Tris Base, 0.1% SDS, 1 mM EDTA, pH 7.7, 0.01-0.09% N,N-dimethylformamide |
For Research Use Only. Not for use in diagnostic procedures.