missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Sample Reducing Agent (10X)

Product Code. 11569166
Click to view available options
Quantity:
250μL
10mL
Unit Size:
10mL
250µL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11569166

Brand: Invitrogen™ NP0009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

NuPAGE Sample Reducing Agent (500mM dithiothreitol)

Ready-to-use reagent to reduce protein samples for protein gel electrophoresis

NuPAGE Sample Reducing Agent contains 500mM dithiothreitol (DTT) at a ready-to-use, 10X concentration in a stabilized liquid form. Use NuPAGE Sample Reducing Agent when running proteins on NuPAGE Novex Bis-Tris gels to maintain protein samples in a reduced state during electrophoresis.

1D Gel Electrophoresis, 2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage NuPAGE™ Sample Reducing Agent (500 mM dithiothreitol) should be stored at 4°C. Guaranteed stable for 6 months unless otherwise specified in the product literature.
Concentration 500 mM
Product Type Sample Reducing Agent
Product Line NuPAGE
Quantity 10mL
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.