missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Transfer Buffer (20X)

Product Code. 10564613
Click to view available options
Quantity:
125mL
1L
Unit Size:
1 L
125mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10564613

Brand: Invitrogen™ NP00061

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Use to transfer proteins from NuPAGE Novex gels to membranes for Western blotting

NuPAGE Transfer Buffer can be used with the Novex Semi-Dry Blotter for semi-dry transfer or with the XCell II™ Blot Module for wet (tank) transfer. NuPAGE Antioxidant may be used with NuPAGE Transfer Buffer to enhance transfer of reduced proteins to membranes.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature. Guaranteed stable for 6 months unless otherwise specified in the product literature.
Buffer Transfer Buffers
Concentration 20 X
Gel Type NuPAGE
Product Type Transfer Buffer
Gel Compatibility NuPAGE™ Gels
Product Line NuPAGE
Quantity 1L
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.