Learn More
Invitrogen™ NuPAGE™ Tris-Acetate SDS Buffer Kit (for Tris-Acetate Gels), Contains 1 ea. LA0041, NP0004, NP0005, NP0007
Product Code. 10718234
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
For Research Use Only. All usage must comply with product instructions.
Product Code. 10718234
Brand: Invitrogen™ LA0050
Edge
For Research Use Only. All usage must comply with product instructions.
Includes
20X NuPAGE Tris-Acetate SDS Running Buffer (500mL), 10X NuPAGE Sample Reducing Agent (250μl), NuPAGE Antioxidant (15mL), 4X NuPAGE LDS Sample Buffer (10mL)
Provide separation of large molecular weight proteins when used with NuPAGE Tris-Acetate SDS Running Buffer
  • NuPAGE Novex Tris-Acetate gels can also be run with Tris-Glycine Native Running Buffer to resolve native proteins more effectively than Tris-Glycine gel system
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Product Type Buffer Kit
Content And Storage NuPAGE™ Tris-Acetate SDS Buffer Kit (for Tris-Acetate Gels) contains:
• 20X NuPAGE™ Tris-Acetate SDS Running Buffer (500 ml): Store at 4°C to 25°C
• 10X NuPAGE™ Sample Reducing Agent (250 μl): Store at 2°C to 8°C
• NuPAGE™ Antioxidant (15 ml): Store at 4°C
• 4X NuPAGE™ LDS Sample Buffer (10 ml): Store at 4°C to 25°C
Gel Type SDS PAGE Gel
Gel Compatibility NuPAGE Tris-Acetate Gel
Product Line NuPAGE
Quantity 1 kit