Learn More
Invitrogen™ NuPAGE™ Tris-Acetate SDS Running Buffer (20X)
Formulated for separation of proteins, in their denatured state, on NuPAGE Novex Tris-Acetate gels
Brand: Invitrogen™ LA0041
69.26 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Requires : The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.
This item is not returnable. View return policy
Description
- NuPAGE Novex Tris-Acetate gels provide excellent separation of large molecular weight proteins when used with NuPAGE Tris-Acetate SDS Running Buffer
- Premixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Specifications
Store at +4°C to 25°C. Guaranteed stable for 6 months unless otherwise specified in the product literature. | |
20 X | |
SDS Running Buffer | |
NuPAGE™ | |
Approved for shipment at Room Temperature or on Wet Ice |
Running Buffers | |
SDS-PAGE | |
NuPAGE™ Tris-Acetate Gels | |
500 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.