Learn More
Invitrogen™ Novex™ pH 3-10 IEF Buffer Kit,Includes LC5300, LC5310, LC5311
Optimized for using with Novex™ pH 3-10 IEF gels
Brand:  Invitrogen™ LC5317
111.47 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10297142
  • £157.00 / 1 set
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Includes: Each kit includes Novex IEF Anode Buffer, 50X (LC5300), Novex IEF Cathode Buffer pH 3-10, 10X (LC5310), Novex IEF Sample Buffer pH 3-10, 2X (LC5311)
Description
Description
  • Concentration: 20X
Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Anode & Cathode Buffers
IEF
IEF Buffer Kit
Novex™
20 X
3 to 10
Novex™ IEF Gels
1 kit