Learn More
Invitrogen™ EGFR (Full-length) Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ KHR9061
Code : 50
Additional Details : Weight : 0.70000kg
Includes: EGFR Antibody-Coated 96-Well Plate; EGFR (Full-length) Standard; Standard Diluent Buffer; EGFR (Full-length) Detection Antibody; anti-rabbit IgG-HRP (100X); HRP Diluent; Wash Buffer Concentrate (25X); Stabilized Chromogen, TMB; Stop Solution; Plate Covers; Detailed protocol with validation tests
Description
Kit is designed to detect and quantify the level of full length EGFR (non-truncated) in fresh or frozen human cell lysates. The assay recognizes both natural and recombinant full length human EGFR, independent of its phosphorylation state. Principle of the method A monoclonal capture antibody specific for EGFR (full-length) has been coated onto the wells of the 96-well plate. During the first incubation, standards of known content and unknown samples are pipetted into the wells and the antigen binds to the immobilized (capture) antibody. After washing, a rabbit antibody specific for the target protein is added to the wells and serves as a detection antibody by binding to the immobilized protein captured during the first incubation. After washing, a horseradish peroxidase labeled anti-rabbit IgG is added. This binds to the detection antibody to complete the four member sandwich. After a third incubation and washing to remove all the unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of target protein present in the original specimen and the optical density can be read on a standard microplate reader. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. Multiple alternatively spliced transcript variants that encode different protein isoforms have been found for this gene.Order Info
Shipping condition: Wet ice
Specifications
P00533 | |
<0.3 ng/mL | |
HRP | |
Cell Lysates | |
Human | |
1956 | |
4 hr. | |
7.1% | |
HRP | |
96 Tests | |
Cell Lysate,10 μL | |
Human | |
4 hr. |
0.31-20 ng/mL | |
0.16 to 10ng/mL | |
ELISA Kit | |
Receptor Tyrosine Kinases | |
Colorimetric Microplate Reader | |
ERBB,ERBB1,HER1,NISBD2,PIG61,mENA | |
8.2% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Detection Antibody, anti-rabbit-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
EGFR | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
For Research Use Only. Not for use in diagnostic procedures.