Learn More
Invitrogen™ Novex™ Power Supply Adapters
The Novex Power Supply Adapters allow users to connect the covered power leads of electrophoresis units to certain power supplies.
Brand: Invitrogen™ ZA10001
14.36 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Includes: One set of adapters and one Allen wrench
Description
The Novex Power Supply Adapters allow you to connect the covered power leads of electrophoresis units to certain power supplies (for example, some power supplies from Bio-Rad Laboratories, Thermo Fisher Scientific).
The following electrophoresis units from Invitrogen have covered power leads to be in compliance with the latest IEC (International Electrotechnical Commission) and EN (European) standards, and reduce the risk of electrical shock due to exposure to bare metal.
• ZOOM IPGRunner Mini-Cell and ZOOM IEF Fractionator
• XCell4 SureLock Midi-Cell
• Mini Gel Tank
• XCell SureLock Mini-Cell (has retractable power leads)
If you are using any of the above electrophoresis units with power supplies that are not designed for use with covered or retractable power leads, you need to use Novex Power Supply Adapters.
Order Info
Shipping Condition: Room temperature
Specifications
Contains 1 set of adapters and 1 Allen wrench | |
Novex | |
Room Temperature |
XCell SureLock Mini, ZOOM IPGRunner Mini, XCell4 SureLock Midi, ZOOM IEF Fractionator, Mini Gel Tank | |
1 set |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.