Learn More
Invitrogen™ ProBond™ Purification System
Designed for the purification of recombinant proteins that contain a polyhistidine (6xHis) sequence
Brand: Invitrogen™ K85001
Code : 50
Additional Details : Weight : 1.06000kg
Includes: 6 x 2mL resin columns and buffers for native and denaturing purification; 12mL ProBond pre-charged resin
Description
- Utilizes proprietary ProBond nickel-chelating resin and is supplied with native and denaturing buffers for efficient purification of recombinant proteins under different conditions
His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Specifications
Suspension | |
Six 2-ml resin columns and buffers for native and denaturing purification. Twelve milliliters of ProBond™ pre-charged resin. Store at +4°C. All reagents are guaranteed stable for 6 months when properly stored. | |
ProBond™ |
ProBond™ Purification System | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.