missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Sharp Pre-stained Protein Standard

Product Code. 10640504
Click to view available options
Quantity:
2 x 250 μL
Unit Size:
500µL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10640504

Brand: Invitrogen™ LC5800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Novex Sharp Pre-stained Protein Standards provided as 2 x 250μL (total of 50 applications of 10μL each) of ready-to-use standardmixture Novex Sharp Pre-Stained standard loading buffer consists of 65 mM Tris pH 6.5, 30% glycerol, 2% SDS, and 2.5 mM ETDA

Designed for accurate, easy, and convenient molecular weight estimation of a wide range of molecular weight proteins during SDS-PAGE and Western blotting

  • Accurate - sharp bands for the accurate estimation of molecular weight
  • Unambiguous - each band in the standard is pre-stained with a unique color for easy interpretation of results
  • Comprehensive - a wide range of molecular weight bands consisting of 12 proteins in the range of 3.5 to 260kDa
  • Convenient - a ready-to-use formulation eliminates the need to heat, reduce, or add sample buffer prior to use
  • Adaptable - suitable for most gel types, recommended for use with Novex™ NuPAGE™, Tris-Glycine, and Tricine gels
  • Gel Compatibility: Bolt Bis-Tris Plus Gels, NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels

The Novex Sharp Protein Standard is also available in an unstained format.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Novex™ Sharp Pre-stained Protein Standards are provided as 2 x 250 μL (total of 50 applications of 10 μL each) of ready-to-use standard mixture. The Novex™ Sharp Pre-Stained standard loading buffer consists of 65 mM Tris pH 6.5, 30% glycerol, 2% SDS, and 2.5 mM ETDA.

Store at -20°C.
Detection Method Colorimetric
Number of Markers 12
Product Type Protein Ladder
Ready to Load Yes
Size Range 3.5 to 260 kDa
Stain Type 3 colors: Pink, Yellow, Blue
Gel Compatibility Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels
Molecular Weight (g/mol) 260, 160, 110, 80, 60, 50, 40, 30, 20, 15, 10, 3.5 kDa
Product Line Novex
Quantity 2 x 250 μL
Shipping Condition Approved for shipment on Wet or Dry Ice
System Type Western Blotting, SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.