Learn More
Invitrogen™ SilverQuest™ Silver Staining Kit
Descripción
Includes
This kit contains sufficient reagents to stain 25 minigels. Contains: 250 mL Sensitizer, 25 mL Stainer, 250 mL Developer, 2mL Developer Enhancer, 250 mL Stopper solution, 60 mL Destainer A, 60 mL Destainer B
The SilverQuest™ Silver Staining Kit provides a rapid and easy method to silver stain proteins and DNA in polyacrylamide gels. The SilverQuest™ Silver Staining Kit is specifically designed to provide sensitive silver staining compatible with mass spectrometry analysis.
- High sensitivity with protein detection at sub-nanogram level
- Staining compatible with mass spectrometry analysis
- Rapid and protein bands can be visualized within an hour
- Easy to use, color-coded solutions for easy staining protocol
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Información de órdenes
Shipping Condition: Room Temperature
Especificaciones
Especificaciones
| Content And Storage | This kit contains sufficient reagents to stain 25 minigels Contains: 250 mL Sensitizer 25 mL Stainer 250 mL Developer 2 mL Developer Enhancer 250 mL Stopper solution 60 mL Destainer A 60 mL Destainer B Store kit at room temperature (15°C to 30°C). Stable for 6 months. |
| Detection Location | In-Gel Detection |
| Detection Method | Colorimetric |
| Label or Dye | SilverQuest Silver Stain |
| Product Type | Protein Gel Staining Kit |
| Product Line | SilverQuest |
| Quantity | 1 kit |
| Target Molecule | Protein |
Seguridad y manipulación
- SilverQuest Silver Staining Kit
- Danger
- Oxidising liquid Category 2
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Carcinogenicity Category 1
- Germ cell mutagenicity Category 2
- H319-Causes serious eye irritation.
- H360-May damage fertility or the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P370+P378-In case of fire: Use to extinguish.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Dimethylformamide, Silver nitrate, Formaldehyde, Methyl alcohol, Tetrasodium EDTA
For Research Use Only. Not for use in diagnostic procedures.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.