missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SilverXpress™ Silver Staining Kit

Product Code. 10052662
Click to view available options
Quantity:
1 kit
Unit Size:
625mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10052662

Brand: Invitrogen™ LC6100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

This kit contains sufficient reagents to stain 25 minigels. Contains: 125 mL Sensitizer, 125 mL Stainer A, 125 mL Stainer B, 125 mL Developer, 125 mL Stopper solution

Staining is complete in a little over one hour

The SilverXpress™ Silver Staining Kit is a highly sensitive silver staining kit providing nanogramLevel sensitivity with minimal background. The easy-to-follow protocol requires less total time than standard Coomassie™.

  • Highly sensitive protein staining
  • Crystal clear backgrounds

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains sufficient reagents to stain 25 minigels

Contains:
125 mL Sensitizer
125 mL Stainer A
125 mL Stainer B
125 mL Developer
125 Stopper solution

Store kit at 2°C to 8°C. Stable for 6 months when stored under these conditions.
Detection Location In-Gel Detection
Detection Method Colorimetric
Label or Dye SilverXpress Silver Stain
Product Type Protein Gel Staining Kit
Product Line SilverXpress
Quantity 1 kit
Shipping Condition Room Temperature
Target Molecule Protein
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • SilverXpress Silver Staining Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after single exposure Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Silver nitrate, Ammonium hydroxide, Sodium hydroxide, Glutaraldehyde, Formaldehyde

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.