Learn More
Invitrogen™ StainEase™ Staining Trays
Stains and solutions can be changed without having to handle the gel
Brand: Invitrogen™ NI2400
Code : 50
Additional Details : Weight : 0.00454kg
Includes: StainEase™ Gel Staining Tray comes with two clear polycarbonate containers, two white polycarbonate strainers, two clear polycarbonate lids with handles and waterproof lid labels.
Description
The StainEase Gel Staining Tray is a convenient way to evenly stain mini-gels and prevent gels from breaking. The staining tray assembly includes a perforated strainer insert that holds the gel and drains away the stain, a lid, and an outer container. Easily fits up to four mini-gels.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Contents: StainEase™ Gel Staining Tray comes with two clear polycarbonate containers, two white polycarbonate strainers, two clear polycarbonate lids with handles and waterproof lid labels. | |
Staining Tray | |
StainEase™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.