Learn More
Invitrogen™ Streptavidin Agarose
For isolating biotinylated molecules and complexes with biotinylated components, including antibodies, antibody-antigen complexes, ligands and receptors, nucleic acids, and nucleic acid-protein complexes
Brand:  Invitrogen™ SA10004
Product Code. 11558976
  • £460.00 / 5mL
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Includes: Streptavidin Agarose is supplied as 10mL of a 50% slurry containing 5mL of packed gel bed in 0.01M sodium phosphate (pH 7.2), 0.15M NaCl, and 0.02% sodium azide.
Description
Description
  • Especially useful as an immunoprecipitation reagent for antibodies that do not bind well to protein A
  • The matrix consists of streptavidin covalently attached to cross-linked 4% agarose beads through a 15-atom spacer arm
  • Streptavidin Agarose can bind 3 to 8mg of biotinylated IgG/mL of gel
Immunoprecipitation, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mg/mL
Streptavidin Agarose is supplied as 10 ml of a 50% slurry containing 5 ml of packed gel bed in 0.01 M sodium phosphate (pH 7.2), 0.15 M NaCl, and 0.02% sodium azide. Store at 4°C.
Streptavidin Agarose
5 mL
Liquid Suspension