Learn More
Invitrogen™ Novex™ TBE-Urea Gels, 6%
Novex™ TBE-Urea Gels are denaturing polyacrylamide gels that resolve single-stranded DNA oligos or RNA into sharp, distinct bands.
Brand: Invitrogen™ EC6865BOX
145.60 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Includes: Catalog numbers for gels refer to a single box. One box contains 10 gels. Novex TBE-Urea Gels have a shelf life of between 4 and 8 weeks depending upon gel type.
Description
Novex™ TBE-Urea Gels are denaturing polyacrylamide gels that resolve single-stranded DNA oligos or RNA into sharp, distinct bands. These 6% gels are optimized for the analysis and purification of products ranging from 25–110 bases, making them an ideal choice for synthetic oligo analysis and purification, RNase Protection Assays (RPA), in vitro transcription studies, and northern blot analysis.
Novex TBE-Urea Gels can be stained by silver staining, ethidium bromide, and SYBR™ Green staining techniques after electrophoresis. The compact gels are available in 6%, 10%, or 15% formats and are designed to run on the XCell SureLock™ Mini-Cell.
Advantages of TBE-Urea Gels:
• Contains 7M urea for maximum denaturation
• Excellent resolution for fast size and purity confirmations of DNA or RNA oligos
• High resolution of short single-strand oligonucleotides
• Requires ∼10% sample concentration and volume of large or agarose gels
• Accurate and reproducible results
Formulation
Novex TBE-Urea gels are made with high-purity reagents, including Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, APS, and 7M urea. Strict quality control ensures that gels and buffers are DNase- and RNase-free.
Recommended Buffers
For optimum performance, Novex™ TBE-Urea sample buffers and Novex™ TBE Running Buffer are strongly recommended for use with these gels. For analytical applications, Novex TBE-Urea Sample Buffer is recommended contains urea, the density agent Ficoll™, which yields sharper, straighter bands than conventional density agents, and the tracking dyes Bromophenol Blue and Xylene Cyanol.
Order Info
Shipping Condition: Wet Ice
Specifications
• Catalog number refers to a single box of 10 gels • Novex™ TBE-Urea Gels have a shelf life of 4 to 8 weeks depending upon gel type Store at 4°C. |
|
XCell SureLock Mini-Cell | |
TBE-Urea | |
25 to 110 bp | |
10 Gels/Box | |
Wet Ice |
Novex TBE-Urea Gels, 6%, 10-well | |
6% | |
TBE Gel | |
10-well | |
Single-stranded DNA Oligos/RNA |
Safety and Handling
- 6% TBE-Urea Gels 1.0 mm, 10 Well
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362-Take off contaminated clothing.
Supplemental information
- MIXTURE LIST-Contains : urea