missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMB Chromogen Solution (for ELISA)

Product Code. 10445723
Click to view available options
Quantity:
500 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10445723

Brand: Invitrogen™ 002023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

May be used as a substrate for horseradish peroxidase (HRP) in ELISAs

Use TMB (3,3', 5,5;-tetramethylbenzidine) chromogen solution as supplied. No dilution or further preparation is required. In the presence of peroxidase (HRP, POD), TMB will be converted to a blue color that can be read at OD650 nm.

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C.
Detection Method Colorimetric
Product Type TMB Solution
Substrate TMB
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase) Substrate
Quantity 500 mL
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.