missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TNF alpha ELISA Kit

Product Code. 10632644
Click to view available options
Quantity:
2 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
192 tests
96 tests
Pack of 5
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10632644

Brand: Invitrogen™ KHC3011C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Human TNF-α Antibody Coated 96-Well Plate
Human TNF-α Standard
Standard Diluent Buffer
Human TNF-α Detection Antibody
Streptavidin-HRP
HRP Diluent
Wash Buffer Concentrate (25X)
Stabilized Chromogen, TMB
Stop Solution
Plate Covers
etailed protocol with validation tests

ELISA

The Human Tumor Necrosis Factor alpha (Hu TNFα) ELISA quantitates Hu TNFα in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TNFα. Principle of the method The Human TNFα solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01375
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 1.7 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 7124
Gene Symbol DIF-alpha,TNFA,TNFSF2,TNLG1F,TNF
Incubation Time 4 hr.
Interassay CV 7.5%
Intraassay CV 4.4
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene TNF-α
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Target TNF alpha
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.