Learn More
Invitrogen™ Novex™ Tricine SDS Running Buffer (10X)
Novex Tricine SDS Running Buffer (10X) is formulated for separation of proteins in their denatured state on tricine gels.
Brand: Invitrogen™ LC1675
60.69 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
Novex Tricine SDS Running Buffer (10X) is formulated for separation of proteins in their denatured state on tricine gels. Tricine gels are specifically designed for the resolution of low molecular weight proteins. In this buffer system, tricine substitutes glycine in the running buffer, resulting in more efficient stacking and destacking of low molecular weight proteins and higher resolution of smaller peptides.
Recommended buffers: Use with tricine sample buffer (Cat. No. LC1676) to obtain optimal separation with tricine SDS running buffer.
Order Info
Shipping Condition: Room Temperature
Specifications
Sample Loading Buffer | |
Liquid | |
500 mL |
10X | |
Novex | |
Water |
For Research Use Only. Not for use in diagnostic procedures.