missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tricine Welcome Pack, 16%

Product Code. p-7224318
Click to view available options
Wells:
10-well
12-well
15-well
Unit Size:
1 set
Each
This item is not returnable. View return policy

Product Code. 16586864

Brand: Invitrogen™ EC6695A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • Mini Gel Tank
  • Novex Tricine Mini Gels (2 boxes, 20 gels)
  • Novex Tricine SDS Running Buffer (10x)
  • Novex Tricine SDS Sample Buffer (2X)
  • NuPAGE Sample Reducing Agent (10X)
  • Spectra Multicolor Low Range Protein Ladder

The Novex Tricine Welcome Pack provides all of the necessary Novex Tricine gels, buffers, and reagents you will need to begin using the Mini Gel Tank.

The Novex Tricine Welcome Pack provides all of the necessary Novex Tricine gels, buffers, and reagents you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies being used.

The Novex Tricine Welcome Pack contains:
• Mini Gel Tank (A25977)
• Novex Tricine Mini Gels (2 boxes, 20 gels)
• Novex Tricine SDS Running Buffer, 10X (LC1675)
• Novex Tricine SDS Sample Buffer, 2X (LC1676)
• NuPAGE Sample Reducing Agent, 10X (NP0009)
• Spectra Multicolor Low Range Protein Ladder (26628)

About the Mini Gel Tank
The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies being used.

About Novex Tricine gels
Novex Tricine gels provide separation of low molecular weight proteins and peptides. The Tricine system is a modification of the tris-glycine discontinuous buffer system developed by Schaegger and von Jagow specifically for peptides and low molecular weight proteins. In this system the tricine replaces the glycine in the running buffer, resulting in more efficient stacking and de-stacking of low molecular weight proteins and higher resolution of smaller peptides.

Transfer
For transfer of proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mini Gel Tank
Gel Percentage 16%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tricine
Separation Range 2.5 to 200 kDa
Wells 10-well
Product Line Novex
Quantity 1 Welcome Pk. kit
Shipping Condition Wet Ice
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.