missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine Native Running Buffer (10X)

Product Code. 11539066
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11539066

Brand: Invitrogen™ LC2672

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Requires

For native running conditions, the Tris-Glycine Native Running and Sample Buffers should be used with NuPAGE™ Tris-Acetate gels.

Use with NuPAGE Novex Tris-Acetate gels

Run NuPAGE Novex Tris-Acetate gels with Tris-Glycine Native Running Buffer to resolve native proteins more effectively than Tris-Glycine gel system

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Running Buffer
Concentration 10X
For Use With (Equipment) Novex Tris-Glycine Gels, NuPAGE Tris-Acetate Gels, Novex Tris-Glycine Gels
Physical Form Liquid
Product Line Novex
Quantity 500 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.