Learn More
Invitrogen™ Novex™ Tris-Glycine SDS Buffer Kit
Made with high-purity reagents and are strictly quality controlled
Brand: Invitrogen™ LC2677
35.55 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Code : 50
Additional Details : Weight : 0.72000kg
Requires : The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.
Description
Pre-mixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Novex Tris-Glycine Gel | |
SDS PAGE Gel | |
Buffer Kit |
• 500 mL 10X Tris-Glycine SDS Running Buffer • 20 mL 2X Tris-Glycine SDS Sample Buffer All Novex™ buffers have a 6 month shelf life. Store at +4°C to 25°C, depending upon formulation. |
|
Novex™ | |
1 kit |
For Research Use Only. Not for use in diagnostic procedures.