missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine SDS Running Buffer (10X)

Product Code. 10040173
Click to view available options
Quantity:
500 mL
5 L
4 x 1 L
Unit Size:
4 L
5 L
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10040173

Brand: Invitrogen™ LC26754

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Novex Tris-Glycine SDS Running Buffer (10X), 500mL

Requires

The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.

Formulated for separation of proteins, in their denatured state, on Novex Tris-Glycine gels

Tris-Glycine gels provide reproducible separation of a wide range of proteins into well-resolved bands, depending upon the gel percentage used.

  • Pre-mixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results
  • Made with high-purity reagents and are strictly quality controlled

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Running Buffer
Concentration 10X
For Use With (Equipment) Novex Tris-Glycine Gels
Product Line Novex
Quantity 4 x 1 L

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.