Learn More
Invitrogen™ Novex™ Tris-Glycine SDS Sample Buffer (2X)
Use with Novex Tris-Glycine Gels
Brand: Invitrogen™ LC2676
27.02 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : CAS Number : 53123-88-9 Weight : 0.02000kg
Description
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Sample Loading Buffer | |
Liquid | |
20 mL |
2X | |
Novex |
For Research Use Only. Not for use in diagnostic procedures.