Learn More
Invitrogen™ Tris-Glycine Transfer Buffer (25X)
Made with high-purity reagents and are strictly quality controlled
Brand: Invitrogen™ LC3675
53.20 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : CAS Number : 53123-88-9 Weight : 0.61000kg
Requires : The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.
Description
Tris-Glycine Transfer Buffer (25X) is optimized for western blot transfer applications using Tris-Glycine gels. Premixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Novex Tris-Glycine Gel | |
Tris-Glycine Gel | |
Nitrocellulose, PVDF, Nylon | |
25X |
Store at room temperature. | |
Room Temperature | |
Novex™ | |
Transfer Buffer |
For Research Use Only. Not for use in diagnostic procedures.