missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ Value™ Tris-Glycine Mini Protein Gel, 4 to 20%, 1.0 mm, 10-well
Separation Range: 20 to 200 kd; Separation Type: Denaturing, Native; For Use With: XCell SureLock Mini
Brand: Invitrogen™ XV04200PK20
Includes: 20 precast gels
Description
Features of the Novex Value gels include:- Great balance of value and performance
- Fast run conditions - separate your proteins using constant voltage in less than 60 minutes
- Improved shelf life - store gels for at least 12 months at 4°C
- Usable for native and denaturing protein assays
Services
Up to 12 months
Specifications
Store in refrigerator (2–8°C). Shelf life up to 12 months depending on gel type. |
|
Mini Gel Tank, XCell SureLock Mini-Cell | |
Mini | |
Tris-Glycine | |
Denaturing, Native | |
Protein | |
Denaturing, Native | |
Gel | |
Tris-Glycine | |
Novex Value™ | |
Approved for shipment at Room Temperature or on Wet Ice | |
1.0 mm | |
1.0 mm | |
8 cm | |
8 cm |
Novex™ Value™ 4 to 20% Tris-Glycine Mini Gels, 20 Gels, 0.039 in. Thickness, 8 x 8cm Gel Size, Gel, For Protein Gel Electrophoresis, 12 Months Shelf Life, 2 to 8°C Storage Temperature | |
4 to 20% | |
1.0 mm | |
Gradient Page | |
up to 25 μL | |
20 to 200 kDa | |
10-well | |
Protein Gel Electrophoresis | |
Molecular Weight | |
20/Pk. | |
Store at 4°C. Shelf life is 12 months. | |
0.039 in. | |
1D Well | |
8 cm | |
8 cm |
For Research Use Only. Not for use in diagnostic procedures.