missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Novex™ WedgeWell™ Tris-Glycine XCell SureLock™ Mini-Cell Welcome Pack, 4 to 20%

The Novex Tris-Glycine Welcome Pack provides all of the necessary Novex Tris-glycine gels, buffers, and reagents needed to begin using the XCell SureLock Mini-Cell.

Brand:  Thermo Scientific™ XP0420D

Additional Details : Weight : 2.95700kg

 View more versions of this product

Product Code. 17753734

  • £498.00 / Each
Estimated Shipment: 19-08-2024
to see stock.
Add to basket

Description

Description

The Novex Tris-Glycine Welcome Pack provides all of the necessary Novex Tris-glycine gels, buffers, and reagents needed to begin using the XCell SureLock Mini-Cell. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.

The Novex Tris-Glycine Welcome Pack contains:
• XCell SureLock Mini-Cell (EI0001)
• Novex Tris-Glycine Mini Gels, WedgeWell format (2 boxes, 20 gels)
• Novex Tris-Glycine SDS Running Buffer, 10X (LC2675)
• Novex Tris-Glycine SDS Sample Buffer, 2X (LC2676)
• NuPAGE Sample Reducing Agent, 10X (NP0004)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)

About the XCell SureLock Mini-Cell 
The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.

About Novex Tris-Glycine gels, WedgeWell format
Novex Tris-Glycine mini gels are polyacrylamide gels based on traditional Laemmli protein electrophoresis, allowing the use of Laemmli sample and running buffers. They enable consistent performance and separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For transferring your proteins to a membrane, we recommend using for Novex Tris-Glycine Native Running Buffer (LC2672) or Novex Tris-Glycine Transfer Buffer (LC3675) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Specifications

Specifications

XCell SureLock Mini-Cell
Mini
Tris-Glycine
10-well
1 Welcome Pk.
4 to 20%
1.0 mm
20 to 200 kDa
Novex™, WedgeWell™
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.