Learn More
Invitrogen™ XCell SureLock™ Mini-Cell
Quickly and easily run up to two Novex 8 x 8cm minigels, leak-free without any clamps or grease
Brand: Invitrogen™ EI0001
571.20 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
The XCell SureLock Mini-Cell incorporates a gel tension wedge, in place of the rear wedge used on earlier models. When the lever on the gel tension wedge is pushed forward into the locked position, an even, horizontal force is generated. This seals the gel/buffer core assembly firmly into position in the lower buffer chamber. The positive locking action of the gel tension wedge ensures a trouble-free, leak-free gel run every time.
- Can be easily set up in just three simple steps
- Available separately or as a part of a complete package
Upgrade XCell II to an XCell SureLock Mini-Cell
- Conversion of an XCell II into an XCell SureLock is easy with the SureLock Retrofit Kit (available separately)
- The kit consists of a SureLock Mini-Cell lid, gel tension wedge, and molded minigel buffer dam
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature

Specifications
Store at room temperature. Warrantied for one year from the time of purchasing. | |
XCell SureLock™ Mini | |
Mini (8 x 8 cm) | |
Mini-Cell Mini-Vertical Electrophoresis System | |
Room Temperature |
2 Mini-gels | |
Novex™ Mini Gels, NuPAGE™ Gels | |
XCell SureLock | |
1 unit | |
500 VDC |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.