Learn More
Invitrogen™ XCell II™ Blot Module
Description
Includes
Anode core, cathode core, sponge pads (8/pack), and instruction manual.
Fits neatly into the XCell SureLock™ and XCell II™ Mini-Cells in place of the gel/buffer core assembly. It requires less than 200mL of transfer buffer for Western, Southern, and northern transfers. Tough platinized titanium and stainless steel electrodes create a uniform electrical field without clamps or hinged gel holders. Maximum blot size is 9 x 9cm.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Product Type | Blot Module |
| Capacity | Up to 2 mini-gels |
| Content And Storage | The XCell II™ Blot Module includes anode core, cathode core, sponge pads (8/pack), and instruction manual. |
| For Use With (Equipment) | XCell SureLock™ Mini and Mini Gel Tank |
| Gel Size | Mini (8 cm x 8 cm) |
| Mode of Transfer | Wet |
| Running Dimension | Vertical |
| Shipping Condition | Room Temperature |
| Gel Compatibility | Bolt Bis-Tris Plus Gel, NuPAGE Gel, Novex Mini Gel |
| Product Line | XCell II |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
