Learn More
Invitrogen™ Buffer Dam for XCell SureLock™ and XCell II™ Blot Module
XCell SureLock and XCell II Blot Module
Brand: Invitrogen™ EI0012
Code : 50
Additional Details : Weight : 0.00454kg
Description
Applications:
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Store at room temperature. | |
Mini | |
Buffer Dam |
XCell SureLock™ Mini | |
XCell II, XCell SureLock | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.