Learn More
Invitrogen™ Platinum Wire for the XCell SureLock™ apparatus (12 inch x 2)
For the XCell SureLock Mini Blot Module
Brand: Invitrogen™ EI9022
Code : 50
Additional Details : Weight : 0.56000kg
Description
Applications:
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Store at room temperature. | |
XCell SureLock™ Mini | |
XCell SureLock | |
Room Temperature |
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis | |
Mini | |
Platinum Wire |
For Research Use Only. Not for use in diagnostic procedures.