Learn More
Invitrogen™ Buffer Core w/ Electrodes for XCell SureLock™ & XCell II™ Blot Module
For the XCell SureLock Mini Blot Module
Brand: Invitrogen™ EI9014
208.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 50
Additional Details : Weight : 0.56000kg
Description
Applications:
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Store at room temperature. | |
XCell SureLock™ Mini | |
XCell II, XCell SureLock | |
Room Temperature |
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis | |
Mini | |
Buffer Core |
For Research Use Only. Not for use in diagnostic procedures.