Learn More
Invitrogen™ Gel Knife
For the XCell SureLock Mini Blot Module
Brand: Invitrogen™ EI9010
Code : 50
Additional Details : Weight : 0.00454kg
Description
Applications:
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Store at room temperature. | |
Universal | |
Novex, XCell SureLock | |
Room Temperature |
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis | |
Mini | |
Cassette Opener |
For Research Use Only. Not for use in diagnostic procedures.