missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZOOM™ Carrier Ampholytes pH 3-10

Product Code. 11500906
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11500906

Brand: Invitrogen™ ZM0021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Small, soluble molecules with both positive and negative charge groups

Use Carrier Ampholytes with the ZOOM IPGRunner™ System which consists of three main components: ZOOM IPGRunner Mini-Cell, ZOOM IPGRunner Cassettes and 7cm ZOOM Strips.

  • ZOOM Carrier Ampholytes sort based on their isoelectric points in an electric field, thereby establishing a pH gradient in solution or contributing to the gradient when used with IPG strips
  • Carrier ampholytes help stabilize the pH gradient and current in IPG strips and aid in protein solubility, resulting in reproducible IEF resolution

2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The ZOOM™ IPGRunner™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini- Cell chamber. The ZOOM™ IPGRunner™ Combo Kit includes the ZOOM™ IPGRunner™ Mini-Cell, electrode wicks, sealing tape, 10 ZOOM™ IPGRunner™ Cassettes, and 12 ZOOM™ Strips, pH 3-10 NL. The ZOOM™ IPGRunner™ Retrofit Kit for XCell SureLock™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid. The ZOOM™ IPGRunner™ Cassettes include electrode wicks and sealing tape.
pH Range pH 3-10
Product Type Carrier Ampholyte
Product Line ZOOM
Quantity 10 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.